Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253737.1 | NP_001240666.1 | 8, 9 | 573 | ||
NM_001253739.1 | NP_001240668.1 | 8, 9 | 584 | |||
NM_001253743.1 | NP_001240672.1 | 9, 10 | 633 | |||
NM_001253745.1 | NP_001240674.1 | 7, 8 | 510 | |||
NM_001253746.1 | NP_001240675.1 | 7, 8 | 510 | |||
NM_001253747.1 | NP_001240676.1 | 8, 9 | 594 | |||
NM_013517.4 | NP_038545.1 | 9, 10 | 714 | |||
GenBank mRNA | AK156654.1 | - | 9, 10 | 717 | ||
AK172591.1 | - | 9, 10 | 717 | |||
AY069980.1 | - | 7, 8 | 476 | |||
AY069981.1 | - | 7, 8 | 476 | |||
BC119164.1 | - | 8, 9 | 581 | 8733913 | ||
BC119190.1 | - | 8, 9 | 581 | 8733939 | ||
M34163.1 | - | 9, 10 | 716 | |||
M99371.1 | - | 9, 10 | 706 | |||
S67072.1 | - | 8, 9 | 565 | |||
S67073.1 | - | 7, 8 | 445 | |||
S67074.1 | - | 8, 9 | 484 | |||
X64223.1 | - | 8, 9 | 573 | |||
X64224.1 | - | 8, 9 | 584 |