Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253737.1 | NP_001240666.1 | 8 | 505 | ||
NM_001253739.1 | NP_001240668.1 | 8 | 516 | |||
NM_001253743.1 | NP_001240672.1 | 9 | 565 | |||
NM_001253745.1 | NP_001240674.1 | 7 | 442 | |||
NM_001253746.1 | NP_001240675.1 | 7 | 442 | |||
NM_001253747.1 | NP_001240676.1 | 8 | 526 | |||
NM_013517.4 | NP_038545.1 | 9 | 646 | |||
GenBank mRNA | AK156654.1 | - | 9 | 649 | ||
AK172591.1 | - | 9 | 649 | |||
AY069980.1 | - | 7 | 408 | |||
AY069981.1 | - | 7 | 408 | |||
BC119164.1 | - | 8 | 513 | 8733913 | ||
BC119190.1 | - | 8 | 513 | 8733939 | ||
M34163.1 | - | 9 | 648 | |||
M99371.1 | - | 9 | 638 | |||
S67072.1 | - | 8 | 497 | |||
S67073.1 | - | 7 | 377 | |||
S67074.1 | - | 8 | 416 | |||
X64223.1 | - | 8 | 505 | |||
X64224.1 | - | 8 | 516 |