Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301412.1 | NP_001288341.1 | 21 | 3194 | ||
NM_008017.4 | NP_032043.3 | 21 | 3118 | |||
GenBank mRNA | AJ534939.1 | - | 20 | 2949 | ||
BC071232.1 | - | 6 | 853 | |||
BC094380.1 | - | 21 | 3090 | 30543190 | ||
U42385.1 | - | 4 | 561 |