Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130412.1 | NP_001123884.1 | 17, 18 | 2357 | ||
NM_015763.4 | NP_056578.2 | 17, 18 | 2312 | |||
NM_172950.3 | NP_766538.2 | 16, 17 | 2213 | |||
GenBank mRNA | AB093214.1 | - | 12, 13 | 1532 | ||
AK014526.1 | - | 16, 17 | 2273 | |||
AK019539.1 | - | 17, 18 | 2450 | |||
AK030921.1 | - | 16, 17 | 2216 | |||
AK154837.1 | - | 17, 18 | 2354 | |||
BC042462.1 | - | 16, 17 | 2266 | 4211202 |