Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010229.2 | NP_034359.2 | 9 | 1340 | ||
GenBank mRNA | AK045865.1 | - | 9 | 1163 | ||
AK149292.1 | - | 9 | 1158 | |||
AK163640.1 | - | 9 | 1341 | |||
BC109003.1 | - | 9 | 1133 | |||
BC109004.1 | - | 9 | 1145 | |||
M64689.1 | - | 9 | 1146 | |||
X59398.1 | - | 9 | 1197 |