Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008029.3 | NP_032055.1 | 30 | 4167 | ||
GenBank mRNA | BC138346.1 | - | 30 | 4139 | 8861366 | |
BC138348.1 | - | 30 | 4139 | 8861368 | ||
L07296.1 | - | 30 | 4120 |