Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080855.2 | NP_543131.1 | 5 | 564 | ||
GenBank mRNA | AB030244.1 | - | 5 | 575 | ||
BC132449.1 | - | 5 | 562 | 40130726 | ||
BC137853.1 | - | 5 | 562 | 8860872 | ||
CB518791.1 | - | 3 | 210 | |||
CJ102007.1 | - | 5 | 407 |