Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_126166.4 | NP_569054.2 | 4 | 510 | ||
GenBank mRNA | AF355152.1 | - | 2 | 192 | ||
AF420279.1 | - | 1 | 149 | |||
AK083977.1 | - | 4 | 511 | |||
AK087739.1 | - | 3 | 332 | |||
AK166177.1 | - | 3 | 385 | |||
AK171438.1 | - | 4 | 493 | |||
BC099937.1 | - | 4 | 548 | 30084049 |