Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6pc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289856.1 | NP_001276785.1 | 4 | 401 | ||
NM_021331.4 | NP_067306.1 | 4 | 621 | |||
GenBank mRNA | AK148465.1 | - | 4 | 573 | ||
BQ557220.1 | - | 4 | 401 | |||
Z47787.1 | - | 4 | 599 |