Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271467.1 | NP_001258396.1 | 8 | 1047 | ||
NM_001271468.1 | NP_001258397.1 | 7 | 952 | |||
NM_001271470.1 | NP_001258399.1 | 8 | 1114 | |||
NM_010249.2 | NP_034379.1 | 7 | 952 | |||
NM_207669.2 | NP_997552.1 | 7 | 949 | |||
GenBank mRNA | AK052828.1 | - | 928 | |||
AK139710.1 | - | 8 | 1114 | |||
AK140222.1 | - | 8 | 1000 | |||
AK140907.1 | - | 7 | 860 | |||
AK216963.1 | - | 2 | 75 | |||
AY282800.1 | - | 4 | 409 | |||
AY282802.1 | - | 5 | 604 | |||
AY282803.1 | - | 4 | 409 | |||
AY282804.1 | - | 5 | 491 | |||
AY282805.1 | - | 4 | 492 | |||
BC013558.1 | - | 7 | 843 | 3592119 | ||
CA552206.1 | - | 4 | 369 | |||
M74516.1 | - | 7 | 844 | |||
M74517.1 | - | 7 | 846 |