Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099641.2 | NP_001093111.1 | 2 | 419 | ||
NM_008068.3 | NP_032094.2 | 2 | 419 | |||
GenBank mRNA | AF256197.1 | - | 2 | 115 | ||
AF256198.1 | - | 2 | 115 | |||
AK135145.1 | - | 2 | 399 | |||
AK137331.1 | - | 2 | 399 | |||
BC145158.1 | - | 2 | 137 | 9053709 | ||
BC145702.1 | - | 2 | 137 | 40131005 | ||
BF471204.1 | - | 2 | 306 | |||
X51986.1 | - | 2 | 355 |