Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038701.2 | NP_001033790.1 | 9 | 1144 | ||
NM_008071.3 | NP_032097.1 | 9 | 1196 | |||
GenBank mRNA | AK039070.1 | - | 9 | 1166 | ||
AK083051.1 | - | 9 | 1208 | |||
AK083107.1 | - | 9 | 1145 | |||
AK140718.1 | - | 5 | 739 | |||
AK147247.1 | - | 9 | 1150 | |||
U14420.1 | - | 9 | 1177 |