Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040189.1 | - | 2 | 847 | ||
AK077475.1 | - | 2 | 202 | |||
AK154760.1 | - | 2 | 363 | |||
AK165478.1 | - | 1 | 2445 | |||
BC065140.1 | - | 2 | 236 | |||
BC086671.1 | - | 2 | 240 | 30635559 | ||
D38557.1 | - | 2 | 310 |