Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088379.1 | - | 10 | 1461 | ||
AK134525.1 | - | 8 | 851 | |||
AK163544.1 | - | 8 | 883 | |||
AV027352.2 | - | 5 | 635 | |||
BC010985.1 | - | 11 | 1202 | 4038194 | ||
M96265.1 | - | 10 | 1291 |