Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109657.3 | NP_001103127.1 | 5 | 633 | ||
NM_001277079.1 | NP_001264008.1 | 5 | 462 | |||
NM_001277080.1 | NP_001264009.1 | 4 | 376 | |||
NM_008088.5 | NP_032114.2 | 5 | 439 | |||
GenBank mRNA | AF133184.1 | - | 1 | 311 | ||
AK017394.1 | - | 5 | 433 | |||
AK076926.1 | - | 6 | 515 | |||
AK152149.1 | - | 3 | 136 | |||
AK154463.1 | - | 5 | 436 | |||
AK172943.1 | - | 1 | 255 | |||
BC120528.1 | - | 5 | 360 | 40129451 | ||
BC120530.1 | - | 5 | 360 | 40129453 | ||
BU053145.1 | - | 5 | 443 | |||
CJ146254.1 | - | 4 | 368 | |||
U19860.1 | - | 5 | 465 |