Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301628.1 | NP_001288557.1 | 15, 16 | 2245 | ||
NM_027604.4 | NP_081880.2 | 16, 17 | 2332 | |||
GenBank mRNA | AF468037.1 | - | 16, 17 | 2236 | ||
AK014891.1 | - | 10, 11 | 1543 | |||
AK122301.1 | - | 15, 16 | 2160 | |||
AK133852.1 | - | 15, 16 | 2154 | |||
AK145749.1 | - | 16, 17 | 2236 | |||
AK161469.1 | - | 16, 17 | 2275 | |||
AK161771.1 | - | 1, 2 | 203 | |||
AK163315.1 | - | 15, 16 | 2175 | |||
AK168755.1 | - | 15, 16 | 2180 | |||
BC003978.1 | - | 3, 4 | 591 | |||
BC050042.1 | - | 16, 17 | 2248 | 30022307 |