Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038010.2 | NP_001033099.1 | 12 | 1837 | ||
NM_020004.5 | NP_064388.2 | 12 | 1840 | |||
GenBank mRNA | AF254441.1 | - | 12 | 1777 | ||
AK137358.1 | - | 12 | 1817 | |||
AK145205.1 | - | 10 | 1837 | |||
AK147042.1 | - | 12 | 1785 | |||
AK150225.1 | - | 12 | 1818 | |||
AK154866.1 | - | 12 | 1828 | |||
AK155347.1 | - | 12 | 2014 | |||
AK155791.1 | - | 12 | 1839 | |||
AK155842.1 | - | 12 | 1838 | |||
AK158079.1 | - | 12 | 1828 | |||
AK186107.1 | - | 2 | 103 | |||
BC011088.1 | - | 12 | 1747 | |||
BC051952.1 | - | 12 | 1822 | |||
BC060050.1 | - | 12 | 1822 | |||
BC063752.1 | - | 12 | 1822 | 6333090 |