Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038010.2 | NP_001033099.1 | 6, 7 | 1128 | ||
NM_020004.5 | NP_064388.2 | 6, 7 | 1128 | |||
GenBank mRNA | AF254441.1 | - | 6, 7 | 1065 | ||
AK137358.1 | - | 6, 7 | 1105 | |||
AK145205.1 | - | 5, 6 | 1026 | |||
AK147042.1 | - | 6, 7 | 1076 | |||
AK150225.1 | - | 6, 7 | 1106 | |||
AK154866.1 | - | 6, 7 | 1116 | |||
AK155791.1 | - | 6, 7 | 1130 | |||
AK155842.1 | - | 6, 7 | 1129 | |||
AK158079.1 | - | 6, 7 | 1116 | |||
BC051952.1 | - | 6, 7 | 1110 | |||
BC060050.1 | - | 6, 7 | 1110 | |||
BC063752.1 | - | 6, 7 | 1113 | 6333090 |