Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010269.3 | NP_034399.1 | 10 | 1356 | ||
GenBank mRNA | AK004887.1 | - | 10 | 1302 | ||
AK082958.1 | - | 10 | 1318 | |||
AK146835.1 | - | 10 | 1346 | |||
AK161122.1 | - | 10 | 1255 | |||
AK166799.1 | - | 10 | 1271 | |||
AK166859.1 | - | 10 | 1270 | |||
AK187786.1 | - | 2 | 144 | |||
AK188572.1 | - | 1 | 39 | |||
AK208171.1 | - | 1 | 39 | |||
AK208831.1 | - | 1 | 39 | |||
AK213851.1 | - | 1 | 39 | |||
BC025070.1 | - | 10 | 1265 | 3710712 | ||
Y17851.1 | - | 10 | 1353 |