Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF297615.1 | - | 6, 7 | 693 | ||
AK004753.1 | - | 6, 7 | 756 | |||
AK159087.1 | - | 6, 7 | 765 | |||
BC006810.1 | - | 4, 5 | 446 | 3154188 | ||
M26925.1 | - | 5, 6 | 537 | |||
M85153.1 | - | 6, 7 | 636 |