Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081125.1 | NP_001074594.1 | 14 | 4598 | ||
GenBank mRNA | AK051858.1 | - | 1 | 923 | ||
AK078489.1 | - | 1 | 1847 | |||
BC031171.1 | - | 1 | 1232 | |||
BC082604.1 | - | 14 | 4539 | 30672479 | ||
BC085190.1 | - | 14 | 4514 | 30694633 | ||
X99104.1 | - | 14 | 4358 |