Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010297.2 | NP_034427.2 | 6 | 697 | ||
GenBank mRNA | AF462147.1 | - | 5 | 559 | ||
AK085727.1 | - | 6 | 1120 | |||
BC110630.2 | - | 5 | 559 | 40052661 | ||
DQ109809.1 | - | 6 | 697 |