Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010303.3 | NP_034433.3 | 2, 3 | 694 | ||
GenBank mRNA | AK149766.1 | - | 2, 3 | 588 | ||
BC057665.1 | - | 2, 3 | 548 | 4918717 | ||
M63660.1 | - | 2, 3 | 642 |