Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010306.3 | NP_034436.1 | 3, 4 | 454 | ||
GenBank mRNA | AK004566.1 | - | 3, 4 | 448 | ||
AK077490.1 | - | 3, 4 | 416 | |||
AK167438.1 | - | 3, 4 | 408 | |||
AK168578.1 | - | 3, 4 | 392 | |||
AK169067.1 | - | 3, 4 | 399 | |||
AK169285.1 | - | 3, 4 | 429 | |||
BC020009.1 | - | 3, 4 | 426 | |||
BC041107.1 | - | 3, 4 | 365 | 2648164 | ||
S79610.1 | - | 2, 3 | 179 |