Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013530.1 | NP_038558.1 | 4 | 526 | ||
GenBank mRNA | AK014789.1 | - | 4 | 245 | ||
AK030354.1 | - | 3 | 467 | |||
BC018239.1 | - | 4 | 526 | 4511683 |