Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010324.2 | NP_034454.2 | 9 | 1274 | ||
GenBank mRNA | AK146445.1 | - | 9 | 1176 | ||
AK147807.1 | - | 9 | 1197 | |||
AK147934.1 | - | 9 | 1178 | |||
BC002057.1 | - | 9 | 1162 | 3489628 | ||
J02623.1 | - | 9 | 1178 |