Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Got2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010325.2 | NP_034455.1 | 7 | 813 | ||
GenBank mRNA | AK098166.1 | - | 7 | 839 | ||
AK136556.1 | - | 7 | 850 | |||
AK147953.1 | - | 7 | 853 | |||
AK149886.1 | - | 7 | 850 | |||
AK149926.1 | - | 7 | 850 | |||
AK150194.1 | - | 6 | 569 | |||
AK152921.1 | - | 7 | 850 | |||
AK155075.1 | - | 7 | 844 | |||
AK167767.1 | - | 7 | 839 | |||
BC089015.1 | - | 7 | 839 | 30606379 | ||
BC089341.1 | - | 7 | 851 | 6832287 | ||
J02622.1 | - | 7 | 813 | |||
U82470.1 | - | 7 | 781 |
Gene Symbol: Gm10874 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133132.1 | - | 1 | 1079 |