Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016697.3 | NP_057906.2 | 6 | 1490 | ||
GenBank mRNA | AF185614.1 | - | 6 | 1430 | ||
AK146702.1 | - | 6 | 1467 | |||
AK159394.1 | - | 6 | 1459 | |||
AK159435.1 | - | 6 | 1459 | |||
AK159764.1 | - | 6 | 1467 | |||
AK168402.1 | - | 6 | 1468 | |||
BC036126.1 | - | 6 | 1437 | 4973409 |