Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016697.3 | NP_057906.2 | 2 | 455 | ||
GenBank mRNA | AF185614.1 | - | 2 | 395 | ||
AK077942.1 | - | 2 | 433 | |||
AK146702.1 | - | 2 | 432 | |||
AK159394.1 | - | 2 | 424 | |||
AK159435.1 | - | 2 | 424 | |||
AK159764.1 | - | 2 | 432 | |||
AK168402.1 | - | 2 | 433 | |||
BC036126.1 | - | 2 | 402 | 4973409 |