Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008163.3 | NP_032189.1 | 5 | 757 | ||
GenBank mRNA | AK077750.1 | - | 4 | 577 | ||
AK079087.1 | - | 4 | 585 | |||
AK148148.1 | - | 5 | 722 | |||
AK153557.1 | - | 4 | 690 | |||
AK155800.1 | - | 5 | 712 | |||
AK166142.1 | - | 5 | 711 | |||
AK170845.1 | - | 4 | 548 | |||
AK172002.1 | - | 5 | 733 | |||
AK181485.1 | - | 1 | 50 | |||
AK212651.1 | - | 1 | 82 | |||
AK216513.1 | - | 1 | 82 | |||
AK217799.1 | - | 1 | 82 | |||
BC052377.1 | - | 5 | 757 | 5696248 | ||
BC085254.1 | - | 5 | 593 | 5343274 | ||
D85748.1 | - | 3 | 381 | |||
U07617.1 | - | 4 | 506 |