Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008163.3 | NP_032189.1 | 5 | 698 | ||
GenBank mRNA | AK077750.1 | - | 4 | 518 | ||
AK079087.1 | - | 4 | 526 | |||
AK148148.1 | - | 5 | 663 | |||
AK153557.1 | - | 4 | 631 | |||
AK155800.1 | - | 5 | 653 | |||
AK166142.1 | - | 5 | 652 | |||
AK170845.1 | - | 4 | 489 | |||
AK172002.1 | - | 5 | 674 | |||
AK212651.1 | - | 1 | 23 | |||
AK216513.1 | - | 1 | 23 | |||
AK217799.1 | - | 1 | 23 | |||
BC052377.1 | - | 5 | 698 | 5696248 | ||
BC085254.1 | - | 5 | 534 | 5343274 | ||
D85748.1 | - | 3 | 322 | |||
U07617.1 | - | 4 | 447 |