Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010348.3 | NP_034478.1 | 9 | 1859 | ||
NM_146072.4 | NP_666184.2 | 9 | 1859 | |||
GenBank mRNA | AK042635.1 | - | 9 | 1860 | ||
AK043542.1 | - | 9 | 1839 | |||
AK043737.1 | - | 9 | 1823 | |||
AK044250.1 | - | 9 | 1818 | |||
AK044420.1 | - | 9 | 1814 | |||
AK046122.1 | - | 10 | 1911 | |||
AK048634.1 | - | 9 | 1830 | |||
AK133667.1 | - | 9 | 1845 | |||
AK200858.1 | - | 3 | 354 | |||
BC031822.1 | - | 8 | 1281 | 4501004 | ||
BC049275.1 | - | 8 | 1281 | 4501004 | ||
X66118.1 | - | 9 | 1845 |