Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081097.2 | NP_001074566.1 | 13 | 1935 | ||
GenBank mRNA | AF245444.1 | - | 5 | 730 | ||
BC118004.1 | - | 13 | 1930 | |||
CF744155.1 | - | 4 | 588 |