Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163434.1 | NP_001156906.1 | 9 | 2086 | ||
NM_022310.3 | NP_071705.3 | 8 | 2168 | |||
GenBank mRNA | AJ002387.1 | - | 8 | 1947 | ||
AK004578.1 | - | 8 | 2163 | |||
AK076079.1 | - | 9 | 2082 | |||
AK077463.1 | - | 9 | 2087 | |||
AK144650.1 | - | 9 | 2081 | |||
AK144742.1 | - | 9 | 2084 | |||
AK146647.1 | - | 9 | 2085 | |||
AK148539.1 | - | 9 | 2085 | |||
AK151647.1 | - | 9 | 2084 | |||
AK151806.1 | - | 9 | 2085 | |||
AK152020.1 | - | 9 | 2082 | |||
AK152958.1 | - | 9 | 2085 | |||
AK159719.1 | - | 9 | 2082 | |||
AK160821.1 | - | 2085 | ||||
AK166207.1 | - | 9 | 2083 | |||
AK166739.1 | - | 9 | 2082 | |||
AK167011.1 | - | 9 | 2082 | |||
AK168012.1 | - | 9 | 2086 | |||
AK168230.1 | - | 9 | 2080 | |||
AK169034.1 | - | 9 | 2086 | |||
AK186478.1 | - | 1 | 22 | |||
AK192310.1 | - | 1 | 19 | |||
AK218616.1 | - | 1 | 102 | |||
BC050927.1 | - | 8 | 2145 | 6415084 | ||
D78645.1 | - | 8 | 2028 | |||
M19351.1 | - | 1 | 397 | |||
M30779.1 | - | 1 | 477 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AY351588.1 | - | 213 | Chr. - - [-] on Build |