Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163434.1 | NP_001156906.1 | 3 | 482 | ||
NM_022310.3 | NP_071705.3 | 2 | 564 | |||
GenBank mRNA | AJ002387.1 | - | 2 | 343 | ||
AK004578.1 | - | 2 | 559 | |||
AK076079.1 | - | 3 | 478 | |||
AK077463.1 | - | 3 | 482 | |||
AK144650.1 | - | 3 | 478 | |||
AK144742.1 | - | 3 | 478 | |||
AK146647.1 | - | 3 | 481 | |||
AK148539.1 | - | 3 | 481 | |||
AK149447.1 | - | 3 | 481 | |||
AK151244.1 | - | 2 | 430 | |||
AK151647.1 | - | 3 | 480 | |||
AK151806.1 | - | 3 | 481 | |||
AK152020.1 | - | 3 | 478 | |||
AK152521.1 | - | 2 | 561 | |||
AK152958.1 | - | 3 | 481 | |||
AK159719.1 | - | 3 | 478 | |||
AK160821.1 | - | 482 | ||||
AK166207.1 | - | 3 | 478 | |||
AK166739.1 | - | 3 | 478 | |||
AK167011.1 | - | 3 | 478 | |||
AK168012.1 | - | 3 | 482 | |||
AK168230.1 | - | 3 | 477 | |||
AK169034.1 | - | 3 | 482 | |||
AK178434.1 | - | 1 | 76 | |||
AK178841.1 | - | 1 | 77 | |||
AK181293.1 | - | 1 | 76 | |||
AK185871.1 | - | 1 | 76 | |||
AK188133.1 | - | 1 | 76 | |||
AK188254.1 | - | 1 | 76 | |||
AK192496.1 | - | 1 | 73 | |||
AK195133.1 | - | 1 | 38 | |||
AK197130.1 | - | 1 | 29 | |||
AK198934.1 | - | 1 | 37 | |||
AK200033.1 | - | 1 | 36 | |||
BC005785.1 | - | 1 | 198 | |||
BC050927.1 | - | 2 | 541 | 6415084 | ||
D78645.1 | - | 2 | 424 |