Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291075.1 | NP_001278004.1 | 12, 13 | 1430 | ||
NM_008186.4 | NP_032212.3 | 11, 12 | 1463 | |||
GenBank mRNA | AK005065.1 | - | 11, 12 | 1455 | ||
BC052837.1 | - | 12, 13 | 1442 | 30049274 |