Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008188.2 | NP_032214.1 | 4, 5 | 917 | ||
GenBank mRNA | AK017246.1 | - | 4, 5 | 930 | ||
AK142140.1 | - | 4, 5 | 936 | |||
BC002024.1 | - | 4, 5 | 919 | 3591186 | ||
BC012688.1 | - | 3, 4 | 865 | 3987438 | ||
BC046800.1 | - | 4, 5 | 917 | 6403880 | ||
U83176.1 | - | 4, 5 | 911 |