Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gypa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010369.3 | NP_034499.3 | 7, 8 | 491 | ||
GenBank mRNA | AK145998.1 | - | 7, 8 | 492 | ||
AK157851.1 | - | 7, 8 | 492 | |||
AK162614.1 | - | 7, 8 | 476 | |||
BC148192.1 | - | 7, 8 | 755 | 40045496 |