Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013542.2 | NP_038570.1 | 5 | 761 | ||
GenBank mRNA | AK157856.1 | - | 5 | 743 | ||
AK167770.1 | - | 5 | 724 | |||
BC002085.1 | - | 5 | 758 | 3592898 | ||
CT010272.1 | - | 5 | 650 | |||
M12302.1 | - | 5 | 665 | |||
X04072.1 | - | 5 | 761 |