Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010403.2 | NP_034533.1 | 7 | 1012 | ||
GenBank mRNA | AF104312.1 | - | 7 | 1012 | ||
AK149565.1 | - | 7 | 1019 | |||
BC018587.1 | - | 7 | 990 | |||
BC119535.1 | - | 7 | 1012 | 40056371 | ||
BC119536.1 | - | 7 | 1012 | 40056372 |