Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008214.4 | NP_032240.3 | 5 | 607 | ||
GenBank mRNA | AK011685.1 | - | 5 | 523 | ||
AK088824.1 | - | 5 | 560 | |||
AK146922.1 | - | 5 | 560 | |||
AK150081.1 | - | 5 | 526 | |||
AK152329.1 | - | 5 | 539 | |||
AK166826.1 | - | 5 | 523 | |||
AK166892.1 | - | 5 | 570 | |||
BC020088.1 | - | 5 | 542 | 4008479 | ||
U39473.1 | - | 5 | 587 |