Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008225.2 | NP_032251.2 | 5 | 423 | ||
GenBank mRNA | AK040049.1 | - | 5 | 411 | ||
AK088723.1 | - | 5 | 405 | |||
BC007469.1 | - | 5 | 393 | 3593456 | ||
D42120.1 | - | 5 | 354 | |||
X84797.1 | - | 5 | 391 |