Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008225.2 | NP_032251.2 | 11 | 1094 | ||
GenBank mRNA | AK088723.1 | - | 11 | 1076 | ||
AK132608.1 | - | 2 | 142 | |||
BC007469.1 | - | 11 | 1064 | 3593456 | ||
CA321698.1 | - | 2 | 176 | |||
D42120.1 | - | 11 | 1025 | |||
X84797.1 | - | 11 | 1062 |