Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010411.2 | NP_034541.2 | 3 | 248 | ||
GenBank mRNA | AF074881.1 | - | 3 | 195 | ||
AF074882.1 | - | 3 | 195 | |||
AF125536.1 | - | 3 | 180 | |||
BC139300.1 | - | 3 | 276 | 8862322 | ||
BC139301.1 | - | 3 | 276 | 8862323 | ||
BQ932476.1 | - | 3 | 260 | |||
CJ166955.1 | - | 3 | 251 | |||
JN651901.1 | - | 3 | 178 | |||
JN651902.1 | - | 3 | 178 | |||
JN651903.1 | - | 3 | 178 |