Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010411.2 | NP_034541.2 | 1, 2 | 126 | ||
GenBank mRNA | AF074881.1 | - | 1, 2 | 73 | ||
AF074882.1 | - | 1, 2 | 73 | |||
AF098295.1 | - | 1, 2 | 51 | |||
AF125536.1 | - | 1, 2 | 58 | |||
AK145158.1 | - | 1, 2 | 92 | |||
BC139300.1 | - | 1, 2 | 154 | 8862322 | ||
BC139301.1 | - | 1, 2 | 154 | 8862323 | ||
BQ932476.1 | - | 1, 2 | 138 | |||
CJ166955.1 | - | 1, 2 | 129 | |||
JN651901.1 | - | 1, 2 | 56 | |||
JN651902.1 | - | 1, 2 | 56 | |||
JN651903.1 | - | 1, 2 | 56 | |||
JN651904.1 | - | 1, 2 | 56 |