Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159627.1 | NP_001153099.1 | 6 | 1287 | ||
NM_001159628.1 | NP_001153100.1 | 6 | 1295 | |||
NM_010417.2 | NP_034547.2 | 6 | 1295 | |||
NM_181273.4 | NP_851790.1 | 6 | 1287 | |||
GenBank mRNA | AF082567.1 | - | 6 | 1407 | ||
AK032895.1 | - | 6 | 1207 | |||
AK081330.1 | - | 6 | 1263 | |||
AK086923.1 | - | 6 | 1206 | |||
AK129194.1 | - | 6 | 1243 | |||
BC048237.1 | - | 6 | 1135 | 6474714 | ||
BC054442.1 | - | 6 | 1135 | 6474713 |