Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159328.1 | NP_001152800.1 | 17 | 1673 | ||
NM_008244.3 | NP_032270.3 | 17 | 1670 | |||
GenBank mRNA | AK046299.1 | - | 17 | 1631 | ||
AK145031.1 | - | 17 | 1593 | |||
AK166441.1 | - | 17 | 1644 | |||
AK205770.1 | - | 2 | 96 | |||
AK209356.1 | - | 2 | 96 | |||
AK216327.1 | - | 2 | 96 | |||
BC003239.1 | - | 17 | 1614 | 3497966 | ||
D50050.1 | - | 17 | 1620 |