Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008246.2 | NP_032272.2 | 11 | 1361 | ||
GenBank mRNA | AK004779.1 | - | 11 | 1300 | ||
AK041662.1 | - | 1 | 2648 | |||
AK135865.1 | - | 11 | 1182 | |||
AK158838.1 | - | 11 | 1362 | |||
AK169626.1 | - | 11 | 1301 | |||
AK188731.1 | - | 1 | 72 | |||
BC118047.1 | - | 11 | 1181 | 40106779 | ||
D88315.1 | - | 11 | 1234 |