Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008255.2 | NP_032281.2 | 19 | 2616 | ||
GenBank mRNA | AK079302.1 | - | 7 | 815 | ||
AK159899.1 | - | 18 | 2504 | |||
AK198607.1 | - | 3 | 231 | |||
BC059873.1 | - | 10 | 1297 | |||
BC085083.1 | - | 19 | 2528 | 5320486 | ||
M62766.1 | - | 4 | 478 |