Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008255.2 | NP_032281.2 | 14, 15 | 2030 | ||
GenBank mRNA | AK079302.1 | - | 2, 3 | 229 | ||
AK159899.1 | - | 13, 14 | 1918 | |||
BC019782.1 | - | 13, 14 | 1906 | |||
BC059873.1 | - | 5, 6 | 711 | |||
BC085083.1 | - | 14, 15 | 1942 | 5320486 |